Thermo Scientific Chemicals

Potassium chloride, Spectroscopy Grade, Thermo Scientific Chemicals

Catalog number: 039795.22
100 g, Each
Thermo Scientific Chemicals

Potassium chloride, Spectroscopy Grade, Thermo Scientific Chemicals

Catalog number: 039795.22
100 g, Each
Quantity
Catalog number: 039795.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7447-40-7
IUPAC Name
potassium chloride
Molecular Formula
ClK
InChI Key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES
[Cl-].[K+]
Appearance (Color)
White
Form
Free flowing powder or crystalline powder
Assay (unspecified)
≥99.0%

Description

It is used to replenish electrolytes and restore water electrolyte balance in treating hypokalemia. It is also used in buffers, fertilizers and explosives and finds useful advantages in medicine, scientific applications, and food processing. It plays an important role for the extraction and solubilization of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text