Aluminum oxide, gamma-phase, 99.97% (metals basis), Thermo Scientific Chemicals
Aluminum oxide, gamma-phase, 99.97% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, gamma-phase, 99.97% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039812.14
also known as 039812-14
Price (USD)/ Each
42.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.80 
Save 7.15 (14%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
42.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.80 
Save 7.15 (14%)
Add to cart
Aluminum oxide, gamma-phase, 99.97% (metals basis), Thermo Scientific Chemicals
Catalog number039812.14
Price (USD)/ Each
42.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.80 
Save 7.15 (14%)
-
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.03% max.
Aluminum oxide, is useful to produce aluminum metal. It has many uses in pharmaceutical and industrial manufacturing processes as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements, available in toothpaste, as a dispersing agent food additive, and for uses in hemodialysis. Tailored aluminum oxide layers plays an important role for the bipolar current adjustment in the plasma electrolytic oxidation process. Red phosphorus/aluminum oxide composites play an essential role as flame retardants in recycled poly(ethylene terephthalate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only