Thermo Scientific Chemicals

Scandium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 039996.04
2 g, Under Argon, Each
Thermo Scientific Chemicals

Scandium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 039996.04
2 g, Under Argon, Each
Quantity
Packaging
Catalog number: 039996.04
also known as 039996-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-20-2
IUPAC Name
scandium
Molecular Formula
Sc
InChI Key
SIXSYDAISGFNSX-UHFFFAOYSA-N
SMILES
[Sc]
Total Rare Earth Oxide Impurities
0.1% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text