Osmium powder, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Osmium powder, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Osmium powder, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 039998.06
also known as 039998-06
Price (USD)/ Each
642.65
Online exclusive
714.00 
Save 71.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
642.65
Online exclusive
714.00 
Save 71.35 (10%)
Add to cart
Osmium powder, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number039998.06
Price (USD)/ Each
642.65
Online exclusive
714.00 
Save 71.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-04-2
IUPAC Nameosmium
Molecular FormulaOs
InChI KeySYQBFIAQOQZEGI-UHFFFAOYSA-N
SMILES[Os]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, -20 mesh
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only