Thermo Scientific Chemicals

Terbium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 040032.04
2 g, Each
Thermo Scientific Chemicals

Terbium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 040032.04
2 g, Each
Quantity
Catalog number: 040032.04
also known as 040032-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14284-95-8
IUPAC Name
terbium(3+) tris(2,4-dioxopentan-3-ide)
Molecular Formula
C15H21O6Tb
InChI Key
MAPWZFZLNRMAMO-UHFFFAOYSA-N
SMILES
[Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Appearance (Color)
Yellow
Form
Powder
Assay from Supplier's CofA
Terbium (Tb): Report

Description

Terbium(III) 2,4-pentanedionate is used as an active pharmaceutical ingredient. It is also used as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text