Terbium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals
Terbium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Terbium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 040032.09
also known as 040032-09
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
Add to cart
Terbium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number040032.09
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS14284-95-8
IUPAC Nameterbium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21O6Tb
InChI KeyMAPWZFZLNRMAMO-UHFFFAOYSA-N
SMILES[Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay from Supplier's CofATerbium (Tb): Report
Terbium(III) 2,4-pentanedionate is used as an active pharmaceutical ingredient. It is also used as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only