Thermo Scientific Chemicals

Tantalum(V) isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol/hexane, Thermo Scientific Chemicals

Catalog number: 040038.AE
100 mL, Each
Thermo Scientific Chemicals

Tantalum(V) isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol/hexane, Thermo Scientific Chemicals

Catalog number: 040038.AE
100 mL, Each
Quantity
Catalog number: 040038.AE
also known as 040038-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16761-83-4
IUPAC Name
pentakis(propan-2-ol) tantalum
Molecular Formula
C15H40O5Ta
InChI Key
KWUQLGUXYUKOKE-UHFFFAOYSA-N
SMILES
[Ta].CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O
Total Metal Impurities
0.1% max., 10% w/v in isopropanol/hexane

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text