Thermo Scientific Chemicals

Erbium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 040092.18
50 g, Each
Thermo Scientific Chemicals

Erbium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 040092.18
50 g, Each
Quantity
Catalog number: 040092.18
also known as 040092-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10031-52-4
IUPAC Name
dierbium(3+) octahydrate trisulfate
Molecular Formula
Er2H16O20S3
InChI Key
SMNPRCXUVYGCAE-UHFFFAOYSA-H
SMILES
O.O.O.O.O.O.O.O.[Er+3].[Er+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Erbium(III) sulfate octahydrate is used as a colorant in glass manufacturing and porcelain enamel glazes. High purity Erbium Sulfate is applied as dopant in making optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text