Indium(III) hydroxide, 99.8% (metals basis), Thermo Scientific Chemicals
Indium(III) hydroxide, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium(III) hydroxide, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 040100.18
also known as 040100-18
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
Add to cart
Indium(III) hydroxide, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number040100.18
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS20661-21-6
IUPAC Nameindiganetriol
Molecular FormulaH6InO3
InChI KeyZMFWDTJZHRDHNW-UHFFFAOYSA-N
SMILESO.O.O.[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Indium(III) hydroxide is used as a precursor to indium(III)oxide. It is also applied in batteries, glasses, ceramics, chemical reagents and semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only