Thermo Scientific Chemicals

Indium(III) sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040102.14
25 g, Each
Thermo Scientific Chemicals

Indium(III) sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040102.14
25 g, Each
Quantity
Catalog number: 040102.14
also known as 040102-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13464-82-9
IUPAC Name
diindium(3+) trisulfate
Molecular Formula
In2O12S3
InChI Key
XGCKLPDYTQRDTR-UHFFFAOYSA-H
SMILES
[In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Total Metal Impurities
0.01% max.

Description

Indium(III) sulfate, anhydrous is used in the preparation of indium or indium containing substances like copper indium selenide. It is employed to electroplate indium metal, as a hardening agent in gold electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text