Thermo Scientific Chemicals

Sodium metabisulfite, ACS, 97% min, Thermo Scientific Chemicals

Catalog number: 040115.A4
2.5 kg, Each
Thermo Scientific Chemicals

Sodium metabisulfite, ACS, 97% min, Thermo Scientific Chemicals

Catalog number: 040115.A4
2.5 kg, Each
Quantity
Catalog number: 040115.A4
also known as 040115-A4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7681-57-4
IUPAC Name
disodium (sulfinatooxy)sulfinate
Molecular Formula
Na2O5S2
InChI Key
LDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S(=O)OS([O-])=O
Assay (unspecified)
97.0% Na₂O₅S₂ min
Impurity content
Fe 0.002%
Insoluble material
0.005%
Impurity content
Cl 0.05%
Trace Metals
(as Pb) 0.001%

Description

Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text