Thermo Scientific Chemicals

Indium(III) fluoride, anhydrous, 96%, Thermo Scientific Chemicals

Catalog number: 040118.09
10 g, Each
Thermo Scientific Chemicals

Indium(III) fluoride, anhydrous, 96%, Thermo Scientific Chemicals

Catalog number: 040118.09
10 g, Each
Quantity
Catalog number: 040118.09
also known as 040118-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-52-0
IUPAC Name
indium(3+) trifluoride
Molecular Formula
F3In
InChI Key
JNLSTWIBJFIVHZ-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[In+3]
Assay (unspecified)
95.1% min

Description

Indium(III) fluoride is employed in the synthesis of non-oxide glasses. It promotes chemoselective addition of TMSCN to aldehydes forming the respective cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text