Thermo Scientific Chemicals

Lead(II) carbonate, ACS, Thermo Scientific Chemicals

Catalog number: 040119.36
500 g, Each
Thermo Scientific Chemicals

Lead(II) carbonate, ACS, Thermo Scientific Chemicals

Catalog number: 040119.36
500 g, Each
Quantity
Catalog number: 040119.36
also known as 040119-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
598-63-0
IUPAC Name
λ²-lead(2+) carbonate
Molecular Formula
CO3Pb
InChI Key
MFEVGQHCNVXMER-UHFFFAOYSA-L
SMILES
[Pb++].[O-]C([O-])=O
Impurity content
Nitrate and nitrite (as NO₃) P.T. (limit about 0.005%)
Impurity content
Ca 0.01%
Impurity content
Fe 0.005%
Impurity content
Cl 0.002%
Impurity content
Insoluble in dilute acetic acid 0.02%

Description

Lead(II) carbonate is used in polymerization catalysts. It serves as a curing agent for silicon resins, heat-resistant lubricants, lead-acid storage batteries, wire-reinforced hoses, thermistors and heat stabilizers for polyvinyl chloride polymers. Further, it is used in electrophotography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text