Zirconium(IV) oxide, 18% in H{2}O, colloidal dispersion, stabilized with 1.3% yttrium oxide, Thermo Scientific Chemicals
Zirconium(IV) oxide, 18% in H{2}O, colloidal dispersion, stabilized with 1.3% yttrium oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) oxide, 18% in H{2}O, colloidal dispersion, stabilized with 1.3% yttrium oxide, Thermo Scientific Chemicals

Stabilized with 1.3% yttrium
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040123.36
also known as 040123-36
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1314-23-4
IUPAC Namedioxozirconium
Molecular FormulaO2Zr
InChI KeyMCMNRKCIXSYSNV-UHFFFAOYSA-N
SMILESO=[Zr]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormColloidal dispersion
pH2-4
Particle SizeAverage: 40-60nm (Malvern Zetasizer)
StabilizerYttria, Y₂O₃: 4mol% on solids; 1.4wt% of dispersion
View more
Zirconium(IV) oxide is mainly employed in ceramic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only