Cerium oxide, 20% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
Cerium oxide, 20% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium oxide, 20% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals

Widely applied as a solid electrolyte in fuel cells. Also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
Catalog number 040125.36
also known as 040125-36
Price (USD)/ Each
139.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
139.00
Add to cart
Cerium oxide, 20% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
Catalog number040125.36
Price (USD)/ Each
139.00
-
Add to cart
Chemical Identifiers
CAS1306-38-3
IUPAC Namedioxocerium
Molecular FormulaCeO2
InChI KeyCETPSERCERDGAM-UHFFFAOYSA-N
SMILESO=[Ce]=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH2.5-3.5
FormColloidal dispersion
Assay from Suppliers CofA19.5-21.0%
Assay from Suppliers CofA19.5-21.0%
FormColloidal dispersion
View more
Cerium(IV) oxide is widely applied as a solid electrolyte in fuel cells. It is also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass. It is utilized to increase specific surface area and inhibit sintering as well as pore size growth at a high temperature by addition with CuO/TiO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only