Indium(III) trifluoromethanesulfonate, 99% min, Thermo Scientific Chemicals
Indium(III) trifluoromethanesulfonate, 99% min, Thermo Scientific Chemicals
Thermo Scientific™

Indium(III) trifluoromethanesulfonate, 99% min, Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 040131.18
Price (USD)/ Each
552.65
Online exclusive
614.00 
Save 61.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
552.65
Online exclusive
614.00 
Save 61.35 (10%)
Add to cart
Indium(III) trifluoromethanesulfonate, 99% min, Thermo Scientific Chemicals
Catalog number040131.18
Price (USD)/ Each
552.65
Online exclusive
614.00 
Save 61.35 (10%)
-
Add to cart
Chemical Identifiers
CAS128008-30-0
IUPAC Nameindium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9InO9S3
InChI KeyUCYRAEIHXSVXPV-UHFFFAOYSA-K
SMILES[In+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99% min
Indium(III) trifluoromethanesulfonate is used as a Lewis acid catalyst in organic synthesis and as a co-catalyst in organometallic catalysis. It is also used as a reactant in the preparation of stable indium bacteriochlorins and decahydroquinoline-type toxins through intramolecular hetero Diels-Alder reactions. Further, it is used in the synthesis of benzoxazoles through cyclocondensations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only