Thermo Scientific Chemicals

Copper(II) trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 040133.09
10 g, Each
Thermo Scientific Chemicals

Copper(II) trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 040133.09
10 g, Each
Quantity
Catalog number: 040133.09
also known as 040133-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34946-82-2
IUPAC Name
copper(II) triflate
Molecular Formula
C2H2CuF6O4S2
InChI Key
TXWAGEFXPDWAQH-UHFFFAOYSA-N
SMILES
[Cu].FC(F)(F)S(=O)=O.FC(F)(F)S(=O)=O
Appearance (Color)
White
Assay (special)
≥98.5%
Form
Powder

Description

Copper(II) trifluoromethanesulfonate is a mild lewis acid. It is used as catalyst which promotes dehydration of alcohols and diols to alkenes at ambient temperatures. It is widely used to generate carbenoid species from ?-diazo esters and ketones, via in situ reduction to the Cu(I) species. It is also promotes the reaction between diazo esters and imines to give aziridines. It catalyzes syn-selective aldol condensation of (Z)-silyl enol ethers with aldehydes, Friedel-Crafts alkylation, acylation reactions of aromatics and addition of trimethylsilyl cyanide to carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text