Thermo Scientific Chemicals

Calcium iodate monohydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 040138.22
100 g, Each
Thermo Scientific Chemicals

Calcium iodate monohydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 040138.22
100 g, Each
Quantity
Catalog number: 040138.22
also known as 040138-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10031-32-0
Form
Powder
Assay (unspecified)
≥99.0% (≥61.5% Iodine content)

Description

Calcium iodate monohydrate is used as oxidant added to lotions and ointments, acts as antiseptic and deodorant. It is used in the manufacture of disinfectants and iodine supplement in animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text