Sodium dihydrogen phosphate, 96%, Thermo Scientific Chemicals
Sodium dihydrogen phosphate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium dihydrogen phosphate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 040145.A1
also known as 040145-A1
Price (USD)/ Each
68.65
Online exclusive
76.20 
Save 7.55 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
68.65
Online exclusive
76.20 
Save 7.55 (10%)
Add to cart
Sodium dihydrogen phosphate, 96%, Thermo Scientific Chemicals
Catalog number040145.A1
Price (USD)/ Each
68.65
Online exclusive
76.20 
Save 7.55 (10%)
-
Add to cart
Chemical Identifiers
CAS7558-80-7
IUPAC Namesodium dihydrogen phosphate
Molecular FormulaH2NaO4P
InChI KeyAJPJDKMHJJGVTQ-UHFFFAOYSA-M
SMILES[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95.1% min
Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It was applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only