Thermo Scientific Chemicals

Silicon carbide, alpha-phase, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 040155.36
500 g, Each
Thermo Scientific Chemicals

Silicon carbide, alpha-phase, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 040155.36
500 g, Each
Quantity
Catalog number: 040155.36
also known as 040155-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
409-21-2
IUPAC Name
methanidylidynesilylium
Molecular Formula
CSi
InChI Key
HBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES
[C-]#[Si+]
Total Metal Impurities
0.2% max.

Description

Used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text