Silicon carbide, alpha-phase, 99.8% (metals basis), Thermo Scientific Chemicals
Silicon carbide, alpha-phase, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon carbide, alpha-phase, 99.8% (metals basis), Thermo Scientific Chemicals

Surface Area 9-11 m2/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number 040155.A3
also known as 040155-A3
Price (USD)/ Each
670.00
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
670.00
Add to cart
Silicon carbide, alpha-phase, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number040155.A3
Price (USD)/ Each
670.00
-
Add to cart
Chemical Identifiers
CAS409-21-2
IUPAC Namemethanidylidynesilylium
Molecular FormulaCSi
InChI KeyHBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES[C-]#[Si+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only