Thermo Scientific Chemicals
Graphite plate, pyrolytic, 1.27x9.98x9.98mm (0.05x0.393x.393in), Thermo Scientific Chemicals
Catalog number: 040156.KF
1 Pc., Each
Thermo Scientific Chemicals
Graphite plate, pyrolytic, 1.27x9.98x9.98mm (0.05x0.393x.393in), Thermo Scientific Chemicals
Catalog number: 040156.KF
1 Pc., Each
Quantity
Catalog number: 040156.KF
also known as 040156-KF
Price (USD)
Have Questions?
Appearance (Color)
Grey
Form
Plate, pyrolytic, 1.27(0.05) thick
Length
9.98mm (0.393) length
Width
9.98mm (0.393) width
Description
Graphite plate is used for fuel cells, electrolysis, electroplating, and other various mechanical applications where friction, heat, or electrical properties are factors. It is also used as a solid plate for SEM calibration.