Thermo Scientific Chemicals

Dihydrogen hexachloroplatinate(IV) solution, Pt 20% (cont. Pt), Thermo Scientific Chemicals

Catalog number: 040177.NJ
0.5 g, Each
Thermo Scientific Chemicals

Dihydrogen hexachloroplatinate(IV) solution, Pt 20% (cont. Pt), Thermo Scientific Chemicals

Catalog number: 040177.NJ
0.5 g, Each
Quantity
Catalog number: 040177.NJ
also known as 040177-NJ
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16941-12-1
IUPAC Name
dihydrogen hexachloroplatinumdiuide
Molecular Formula
Cl6H8Pt
InChI Key
QGXPJOUUIMLZTA-UHFFFAOYSA-P
SMILES
[H+].[H+].Cl.Cl.Cl.Cl.Cl.Cl.[Pt--]
Concentration (weight/weight)
Pt 20% min

Description

Dihydrogen hexachloroplatinate(IV) is used as a catalyst in the determination of potassium and in the purification of platinum. It is also used in catalysis reactions, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and in microscopy. In the presence of copper(II) chloride, it is used to catalyze the photooxygenation of alcohols to aldehydes or ketones. It plays an important role in intramolecular hydrosilylation of an alkyne and hydrosilylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text