Search by lot number or partial lot number
Product Type | Specimen Container |
Closure Included | Yes |
Material | Virgin Polypropylene |
Shape | Cylindrical |
Sterility | Non-sterile |
Capacity (Metric) | 40 mL |
Closure Color | White |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Cap and container bagged separately |
Diameter (Metric) | 48 mm |
Label | No Label |
No. per Case | 600/Cs. |
No. per Pack | 600/Pk. |
Unit Size | Case of 600 |
Product Type | Specimen Container |
Closure Included | Yes |
Material | Virgin Polypropylene |
Shape | Cylindrical |
Sterility | Non-sterile |
Capacity (Metric) | 40 mL |
Closure Color | Yellow |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Cap and container bagged separately |
Diameter (Metric) | 48 mm |
Label | Non-tabbed, 10% buffered formalin |
No. per Case | 600/Cs. |
No. per Pack | 600/Pk. |
Unit Size | Case of 600 |
Catalog Number | Specifications | Unit Size | Closure Color | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
040181 also known as 04 0181 | Case of 600 | White | 236.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
040506 also known as 04 0506 | Case of 600 | Yellow | 250.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information: