Thermo Scientific Chemicals

Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 040184.A4
2.5 kg, Each
Thermo Scientific Chemicals

Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 040184.A4
2.5 kg, Each
Quantity
Catalog number: 040184.A4
also known as 040184-A4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1308-06-1
IUPAC Name
dicobalt(3+) λ²-cobalt(2+) tetraoxidandiide
Molecular Formula
Co3O4
InChI Key
UBEWDCMIDFGDOO-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
Total Metal Impurities
0.3% max.

Description

Finds use in enamels, semiconductors, grinding wheels. Used in catalysis, superconductors, ceramics and other fields as an important inorganic material. Also used as catalyst and catalyst carrier, as electrode active material, for glass, porcelain colorants and pigments, as chemical industry oxidant and as a catalyst for organic synthesis, in senior goggles and other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices, enamels, solar energy absorbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text