Thermo Scientific Chemicals

Phosphorus(III) bromide, 99%, Thermo Scientific Chemicals

Catalog number: 040191.14
25 g, Each
Thermo Scientific Chemicals

Phosphorus(III) bromide, 99%, Thermo Scientific Chemicals

Catalog number: 040191.14
25 g, Each
Quantity
Catalog number: 040191.14
also known as 040191-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-60-8
IUPAC Name
tribromophosphane
Molecular Formula
Br3P
InChI Key
IPNPIHIZVLFAFP-UHFFFAOYSA-N
SMILES
BrP(Br)Br
Appearance (Color)
Colorless to pale yellow
Form
Liquid
Assay from Suppliers CofA
≥98.5%

Description

Phosphorus(III) bromide is used mainly for the conversion of alcohols to alkyl bromides and it is used in tests for skin corrosivity. It is used for alkene preparation and it act as a lewis base and a lewis acid. It is used as a catalyst for the alpha-bromination of carboxylic acids and used as intermediates in hell-volhard-zelinsky halogenation. Phosphorus tribromide is used in the manufacture of pharmaceuticals such as alprazolam, methohexital and fenoprofen. It is also used as a fire suppression agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text