Thermo Scientific Chemicals

Ammonium chloride, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 040193.A1
1 kg, Each
Thermo Scientific Chemicals

Ammonium chloride, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 040193.A1
1 kg, Each
Quantity
Catalog number: 040193.A1
also known as 040193-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12125-02-9
IUPAC Name
amine hydrochloride
Molecular Formula
ClH4N
InChI Key
NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES
N.Cl
Residue on ignition
0.01%
Impurity content
Fe 2ppm
Impurity content
SO₄ 0.002%
pH
of a 5% solution 4.5-5.5 at 25?
Heavy Metals
(ICP-OES) 5ppm

Description

Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacturing of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text