Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)
Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)
Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)
Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)
Thermo Scientific Chemicals

Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)

Maximum level of impurities: Insoluble matter 0.01%, Cl 0.001%, PO4 5ppm, SO4 0.003%, NH4 5ppm, Ca 0.002%, Mg 0.001%, Heavy Metals (as Pb) 5ppm, Fe 5ppm, Na 0.03% | CAS: 298-14-6 | CHKO3 | 100.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 040195.22
also known as 040195-22
Price (USD)
34.40
Each
In stock
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
34.40
Each
Add to cart
Chemical Identifiers
CAS298-14-6
IUPAC Namepotassium hydrogen carbonate
Molecular FormulaCHKO3
InChI KeyTYJJADVDDVDEDZ-UHFFFAOYSA-M
SMILES[K+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.7-100.5% KHCO₃
Impurity contentSO₄ 0.003%
Trace Metals(as Pb) 5ppm
Impurity contentCa 0.002%
Impurity contentCl 0.001%
View more
Ask our AI about this Product
Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek's agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
Y26K054Certificate of AnalysisNov 27, 2024040195.36, 040195.22
T30K016Certificate of AnalysisAug 07, 2024040195.36, 040195.22
M25J016Certificate of AnalysisJan 30, 2023040195.36, 040195.22
C02Q01Certificate of AnalysisDec 08, 2021040195.36, 040195.22
N09H035Certificate of AnalysisDec 08, 2021040195.36, 040195.22
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection