Thermo Scientific Chemicals

Cadmium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040203.22
100 g, Each
Thermo Scientific Chemicals

Cadmium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040203.22
100 g, Each
Quantity
Catalog number: 040203.22
also known as 040203-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7790-79-6
Total Metal Impurities
0.01% max.

Description

Cadmium fluoride is used in phosphors, glass and nuclear reactor controls. It is also used in high-temperature lubricants and to start crystals for lasers. It is used in oxygen-sensitive applications such as the production of metallic alloys and it is also used in limited medical treatment protocols. In addition, it can be used in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text