Cerium molybdenum oxide, 99%, Thermo Scientific Chemicals
Cerium molybdenum oxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium molybdenum oxide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 040210.06
also known as 040210-06
Price (USD)/ Each
82.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
82.40
Add to cart
Cerium molybdenum oxide, 99%, Thermo Scientific Chemicals
Catalog number040210.06
Price (USD)/ Each
82.40
-
Add to cart
Chemical Identifiers
CAS13454-70-1
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Cerium molybdenum oxide is used in the aerospace and coating industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only