Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 040216.22
also known as 040216-22
Price (USD)/ Each
453.65
Online exclusive
504.00 
Save 50.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
453.65
Online exclusive
504.00 
Save 50.35 (10%)
Add to cart
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number040216.22
Price (USD)/ Each
453.65
Online exclusive
504.00 
Save 50.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12449-35-3
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystal
Appearance (Color)White
Total Metal Impurities0.005% max.
Ammonium fluoride is used for etching of glass, preservation of food, in printing textile, and as mothproofing agent. It is used in chemical analysis and in brewing. It is used in the synthesis of NaHoF4 and NaEuF4, which are important in solid state lasers as well as photoluminescence. The fluoride ion is used as a base and nucleophile in organic synthesis. Ammonium fluoride providee safe deprotection strategy for the tert-butyldimethylsilyl (TBS) group during RNA synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only