Thermo Scientific Chemicals

Copper(II) tungsten oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 040217.18
50 g, Each
Thermo Scientific Chemicals

Copper(II) tungsten oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 040217.18
50 g, Each
Quantity
Catalog number: 040217.18
also known as 040217-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13587-35-4
IUPAC Name
copper(2+) tungsten tetraoxidandiide
Molecular Formula
CuO4W
InChI Key
FUZOAUOBBKGKBL-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[Cu++].[W]
Total Metal Impurities
0.5% max.

Description

Copper(II) tungsten oxide nanopowder is used for supercapacitor applications. Nano-sized CuWO4 thin films have been fabricated by radio-frequency (R.F.) sputtering deposition, and are used as positive electrode with both LiClO4 liquid electrolyte and LiPON solid electrolyte in rechargeable lithium batteries. Thin films of copper tungstate is used for nitric oxide sensing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text