Thermo Scientific Chemicals

Cobalt(II) bromide hydrate, Thermo Scientific Chemicals

Catalog number: 040225.18
50 g, Each
Thermo Scientific Chemicals

Cobalt(II) bromide hydrate, Thermo Scientific Chemicals

Catalog number: 040225.18
50 g, Each
Quantity
Catalog number: 040225.18
also known as 040225-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85017-77-2
IUPAC Name
λ²-cobalt(2+) dibromide
Molecular Formula
Br2Co
InChI Key
BZRRQSJJPUGBAA-UHFFFAOYSA-L
SMILES
[Co++].[Br-].[Br-]
Assay (unspecified)
Br 61.2% min
Assay (Titration)
99% min anhy

Description

Hygrometers, catalystsCobalt(II) bromide hydrate is mainly used as a catalyst in organic synthesis, like C-metal, C-heteroatom and C-C bond formations, addition to unsaturated bonds, cyclizations, polymerization, oxidations reductions and carbonylations. It reacts with ammonia to form coordination compound bromopentaamminecobalt(III) bromide. It reacts with triphenylphosphine and form complexes of cobalt(II) bromide, which used as a catalyst in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text