Cerium(III) oxalate hydrate, 99%, Thermo Scientific Chemicals
Cerium(III) oxalate hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(III) oxalate hydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 040227.14
also known as 040227-14
Price (USD)/ Each
73.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
73.00
Add to cart
Cerium(III) oxalate hydrate, 99%, Thermo Scientific Chemicals
Catalog number040227.14
Price (USD)/ Each
73.00
-
Add to cart
Chemical Identifiers
CAS15750-47-7
IUPAC Namedicerium(3+) trioxalate
Molecular FormulaC6Ce2O12
InChI KeyZMZNLKYXLARXFY-UHFFFAOYSA-H
SMILES[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
It is used in ceramics, glass, phosphors and pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only