Thermo Scientific Chemicals

Zinc tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040234.09
10 g, Each
Thermo Scientific Chemicals

Zinc tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040234.09
10 g, Each
Quantity
Catalog number: 040234.09
also known as 040234-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13597-56-3
IUPAC Name
zinc(2+) tungsten tetraoxidandiide
Molecular Formula
O4WZn
InChI Key
IGKCQWHBCHMDFW-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[Zn++].[W]
Total Metal Impurities
0.1% max.

Description

It is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text