Selenium(IV) bromide, 99+%, Thermo Scientific Chemicals
Selenium(IV) bromide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Selenium(IV) bromide, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 040240.06
also known as 040240-06
Price (USD)/ Each
57.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
57.90
Add to cart
Selenium(IV) bromide, 99+%, Thermo Scientific Chemicals
Catalog number040240.06
Price (USD)/ Each
57.90
-
Add to cart
Chemical Identifiers
CAS7789-65-3
IUPAC Nametetrabromo-λ⁴-selane
Molecular FormulaBr4Se
InChI KeyVTQZBGAODFEJOW-UHFFFAOYSA-N
SMILESBr[Se](Br)(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Purity≥99.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only