Thermo Scientific Chemicals

Nickel(II) bromide trihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 040242.18
50 g, Each
Thermo Scientific Chemicals

Nickel(II) bromide trihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 040242.18
50 g, Each
Quantity
Catalog number: 040242.18
also known as 040242-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-49-3
IUPAC Name
dibromonickel trihydrate
Molecular Formula
Br2H6NiO3
InChI Key
UQPSGBZICXWIAG-UHFFFAOYSA-L
SMILES
O.O.O.Br[Ni]Br
Assay (unspecified)
97.6% min

Description

It is used in Soporific and sedatives pharmaceutical Industry, Photosensitive industry, Analytical Chemistry, and also electrolyte in high energy battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text