Thermo Scientific Chemicals

Tetraethoxysilane, 99.9%, Thermo Scientific Chemicals

Catalog number: 040251.36
500 g, Each
Thermo Scientific Chemicals

Tetraethoxysilane, 99.9%, Thermo Scientific Chemicals

Catalog number: 040251.36
500 g, Each
Quantity
Catalog number: 040251.36
also known as 040251-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-10-4
IUPAC Name
tetraethyl silicate
Molecular Formula
C8H20O4Si
InChI Key
BOTDANWDWHJENH-UHFFFAOYSA-N
SMILES
CCO[Si](OCC)(OCC)OCC
Assay from Suppliers CofA
≥99.8%
Appearance (Color)
Clear colorless
Form
Liquid

Description

Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text