Thermo Scientific Chemicals

Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals

Catalog number: 040252.14
25 g, Each
Thermo Scientific Chemicals

Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals

Catalog number: 040252.14
25 g, Each
Quantity
Catalog number: 040252.14
also known as 040252-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-97-9
IUPAC Name
dineodymium(3+) trioxidandiide
Molecular Formula
Nd2O3
InChI Key
PLDDOISOJJCEMH-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Nd+3].[Nd+3]
Total Rare Earth Oxide Impurities
1% max.

Description

ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst.Neodymium(III) oxide is an inorganic compound containing inner transition metal neodymium from lanthanides series in +3 oxidation state and oxide in 2:3 ratio. It adopts a hexagonal crystal structure. It is also known as neodymium oxide, neodymiumtrioxide, neodymium(3+)oxide, neodymium sesquioxide and dineodymiumtrioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text