Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals
Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 040252.22
also known as 040252-22
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
Add to cart
Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals
Catalog number040252.22
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1313-97-9
IUPAC Namedineodymium(3+) trioxidandiide
Molecular FormulaNd2O3
InChI KeyPLDDOISOJJCEMH-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Nd+3].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities1% max.
ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst.Neodymium(III) oxide is an inorganic compound containing inner transition metal neodymium from lanthanides series in +3 oxidation state and oxide in 2:3 ratio. It adopts a hexagonal crystal structure. It is also known as neodymium oxide, neodymiumtrioxide, neodymium(3+)oxide, neodymium sesquioxide and dineodymiumtrioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst
RUO – Research Use Only