Thermo Scientific Chemicals

Neodymium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 040253.14
25 g, Each
Thermo Scientific Chemicals

Neodymium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 040253.14
25 g, Each
Quantity
Catalog number: 040253.14
also known as 040253-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13477-91-3
IUPAC Name
dineodymium(3+) octahydrate trisulfate
Molecular Formula
H16Nd2O20S3
InChI Key
IOUARGWNXMWCLD-UHFFFAOYSA-H
SMILES
O.O.O.O.O.O.O.O.[Nd+3].[Nd+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

It is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text