Magnesium oxide substrate, 10x10x0.5mm, polished one side, 111 orientation, Thermo Scientific Chemicals
Magnesium oxide substrate, 10x10x0.5mm, polished one side, 111 orientation, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium oxide substrate, 10x10x0.5mm, polished one side, 111 orientation, Thermo Scientific Chemicals

100 Orientation
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 Ea.
1 Ea.
Catalog number 040273.KU
also known as 040273-KU
Price (USD)/ Each
422.65
Online exclusive
469.00 
Save 46.35 (10%)
-
Add to cart
Quantity:
5 Ea.
Request bulk or custom format
Price (USD)/ Each
422.65
Online exclusive
469.00 
Save 46.35 (10%)
Add to cart
Magnesium oxide substrate, 10x10x0.5mm, polished one side, 111 orientation, Thermo Scientific Chemicals
Catalog number040273.KU
Price (USD)/ Each
422.65
Online exclusive
469.00 
Save 46.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialMagnesium Oxide Substrate
Name NotePolished Two Sides
CAS1309-48-4
ColorWhite
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only