Aluminum oxide substrate, 10x10x1mm, polished one side, C plane, Thermo Scientific Chemicals
Catalog number040281.KT
Price (USD)/ Each
109.00
-
Add to cart
Specifications
Chemical Name or MaterialAluminum Oxide Substrate, C Plane
Name NotePolished One Side
Recommended StorageAmbient temperatures
Quantity1 Ea.
EINECS Number215-691-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.