Dysprosium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Dysprosium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dysprosium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
50 mm
Catalog number 040286.HF
also known as 040286-HF
Price (USD)/ Each
250.00
-
Add to cart
Quantity:
10 mm
Request bulk or custom format
Price (USD)/ Each
250.00
Add to cart
Dysprosium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Catalog number040286.HF
Price (USD)/ Each
250.00
-
Add to cart
Chemical Identifiers
CAS7429-91-6
IUPAC Namedysprosium
Molecular FormulaDy
InChI KeyKBQHZAAAGSGFKK-UHFFFAOYSA-N
SMILES[Dy]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Ta0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only