Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mm
10 mm
50 mm
100 mm
Catalog number 040293.DH
also known as 040293-DH
Price (USD)/ Each
198.00
-
Add to cart
Quantity:
25 mm
Request bulk or custom format
Price (USD)/ Each
198.00
Add to cart
Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Catalog number040293.DH
Price (USD)/ Each
198.00
-
Add to cart
Chemical Identifiers
CAS7439-91-0
IUPAC Namelanthanum
Molecular FormulaLa
InChI KeyFZLIPJUXYLNCLC-UHFFFAOYSA-N
SMILES[La]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Ta0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only