Thermo Scientific Chemicals

Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Catalog number: 040293.DM
100 mm, Each
Thermo Scientific Chemicals

Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Catalog number: 040293.DM
100 mm, Each
Quantity
Catalog number: 040293.DM
also known as 040293-DM
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7439-91-0
IUPAC Name
lanthanum
Molecular Formula
La
InChI Key
FZLIPJUXYLNCLC-UHFFFAOYSA-N
SMILES
[La]
Total Metal Impurities excluding Ta
0.1% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text