Samarium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Samarium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Samarium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mm
50 mm
100 mm
Catalog number 040298.DH
also known as 040298-DH
Price (USD)/ Each
262.00
-
Add to cart
Quantity:
25 mm
Request bulk or custom format
Price (USD)/ Each
262.00
Add to cart
Samarium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Catalog number040298.DH
Price (USD)/ Each
262.00
-
Add to cart
Chemical Identifiers
CAS7440-19-9
IUPAC Namesamarium
Molecular FormulaSm
InChI KeyKZUNJOHGWZRPMI-UHFFFAOYSA-N
SMILES[Sm]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only