Holmium(III) trifluoromethanesulfonate hydrate, 98%, Thermo Scientific Chemicals
Holmium(III) trifluoromethanesulfonate hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium(III) trifluoromethanesulfonate hydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Catalog number 040307.09
also known as 040307-09
Price (USD)/ Each
137.65
Online exclusive
153.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
137.65
Online exclusive
153.00 
Save 15.35 (10%)
Add to cart
Holmium(III) trifluoromethanesulfonate hydrate, 98%, Thermo Scientific Chemicals
Catalog number040307.09
Price (USD)/ Each
137.65
Online exclusive
153.00 
Save 15.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialHolmium(III) trifluoromethanesulfonate hydrate
CAS139177-63-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only