Thermo Scientific Chemicals

Praseodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 040310.09
10 g, Each
Thermo Scientific Chemicals

Praseodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 040310.09
10 g, Each
Quantity
Catalog number: 040310.09
also known as 040310-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52093-27-3
IUPAC Name
praseodymium(3+) tritrifluoromethanesulfonate
Molecular Formula
C3F9O9PrS3
InChI Key
ROUBZIWQWFQCHU-UHFFFAOYSA-K
SMILES
[Pr+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Assay (unspecified)
98% min.
Form
Powder

Description

It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text