Thermo Scientific Chemicals

Terbium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 040312.09
10 g, Each
Thermo Scientific Chemicals

Terbium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 040312.09
10 g, Each
Quantity
Catalog number: 040312.09
also known as 040312-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
148980-31-8
IUPAC Name
terbium(3+) tritrifluoromethanesulfonate
Molecular Formula
C3F9O9S3Tb
InChI Key
JLYAVYSVLLIWEK-UHFFFAOYSA-K
SMILES
[Tb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Appearance (Color)
White
Form
Powder
Assay (unspecified)
Terbium: ≥25.6% (theoretical: 26.2%)

Description

It is used as a reagent & chemical intermediates. It is a raw material used in synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text